Anti-ADAMTS9 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ADAMTS9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-737
- 77837-738
- Antigen symbol:
- ADAMTS9
- ADAMTS9
- Antigen name:
- A disintegrin and metalloproteinase with thrombospondin motifs 9 protein (1276-1506AA)
- A disintegrin and metalloproteinase with thrombospondin motifs 9 protein (1276-1506AA)
- Antigen synonyms:
- ADAMTS9|KIAA1312|A disintegrin and metalloproteinase with thrombospondin motifs 9|ADAM-TS 9|ADAM-TS9|ADAMTS-9|EC 3.4.24.-
- ADAMTS9|KIAA1312|A disintegrin and metalloproteinase with thrombospondin motifs 9|ADAM-TS 9|ADAM-TS9|ADAMTS-9|EC 3.4.24.-
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P2N4
- Q9P2N4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together