Anti-lktA Rabbit Recombinant Polyclonal Antibody (FITC)
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: lktA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mannheimia haemolytica
Spécifications
- Catalog No:
- 77814-905
- 77814-904
- Antigen symbol:
- lktA
- lktA
- Antigen name:
- Recombinant Mannheimia haemolytica Leukotoxin protein (715-953AA)
- Recombinant Mannheimia haemolytica Leukotoxin protein (715-953AA)
- Antigen synonyms:
- lktA|Leukotoxin|Lkt
- lktA|Leukotoxin|Lkt
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Mannheimia haemolytica
- Mannheimia haemolytica
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P0C085
- P0C085
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together