Anti-ZFYVE16 Rabbit Polyclonal Antibody
Catalog # 77804-951
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:FYVE domain containing 16
- Antigen symbol:ZFYVE16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q7Z3T8
- Antigen synonyms:AI035632|B130024H06Rik|B130031L15|DKFZp686E13162|Endofin|Endosomal associated FYVE domain protein|Endosome associated FYVE domain protein|Endosome-associated FYVE domain protein|KIAA0305||mKIAA0305|OTTMUSP00000029589|RGD1564784|ZFY16_HUMAN|ZFYVE16|Zinc finger FYVE domain containing protein 16|Zinc finger FYVE domain-containing protein 16|Zinc finger|FYVE domain containing 16
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ZFYVE16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat