Anti-TRADD Rabbit Polyclonal Antibody
Catalog # 77811-436
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Tumor necrosis factor receptor type 1-associated DEATH domain protein
- Antigen symbol:TRADD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q15628
- Antigen synonyms:9130005N23Rik|AA930854|TNFR1 associated DEATH domain protein|TNFR1-associated DEATH domain protein|TNFRSF1A associated via death domain|TNFRSF1A-associated via death domain|tradd|TRADD_HUMAN|Tumor necrosis factor receptor type 1 associated DEATH domain protein|Tumor necrosis factor receptor type 1-associated DEATH domain protein
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TRADD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat