Anti-PARP9 Rabbit Polyclonal Antibody
77793-249
New Product
- Antigen name:RGD1307534
- Antigen symbol:PARP9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8IXQ6
- Antigen synonyms:ADP-ribosyltransferase diphtheria toxin-like 9|ARTD9|AW214463|B aggressive lymphoma protein|Bagl|BAL|BAL1|BC003281|DKFZp666B0810|DKFZp686M15238|EC 2.4.2.30|FLJ26637|FLJ35310|FLJ41418|FLJ43593|MGC:7868|MGC156739|MGC7868|OTTMUSP00000028034|PARP 9|PARP-9|Parp9|PARP9_HUMAN|Poly (ADP ribose) polymerase family member 9|Poly [ADP-ribose] polymerase 9|RGD1307534
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: PARP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human