Anti-PPP2R2A Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PPP2R2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77795-111
- 77795-112
- Antigen symbol:
- PPP2R2A
- PPP2R2A
- Antigen name:
- Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform protein (2-149AA)
- Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform protein (2-149AA)
- Antigen synonyms:
- 2ABA_HUMAN|Alpha isoform of regulatory subunit B55 protein phosphatase 2|B55A|B55ALPHA|calcineurin|PP2A subunit B B alpha isoform|PP2A subunit B B55 alpha isoform|PP2A subunit B isoform alpha|PP2A subunit B isoform B55-alpha|PP2A subunit B isoform PR55-alpha|PP2A subunit B isoform R2-alpha|PP2A subunit B PR55 alpha isoform|PP2A subunit B R2 alpha isoform|PPP2R2A|PR52A|PR55A|Protein phosphatase 2 (formerly 2A) regulatory subunit B (PR 52) alpha isoform|Protein phosphatase 2 (formerly 2A)|regulatory subunit B (PR52)|alpha isoform|Protein phosphatase 2 regulatory subunit B alpha|Protein phosphatase 2 regulatory subunit B alpha isoform|Protein phosphatase 2 regulatory subunit Balpha|Serine/threonine protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform|Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform|Testicular tissue protein Li 156
- 2ABA_HUMAN|Alpha isoform of regulatory subunit B55 protein phosphatase 2|B55A|B55ALPHA|calcineurin|PP2A subunit B B alpha isoform|PP2A subunit B B55 alpha isoform|PP2A subunit B isoform alpha|PP2A subunit B isoform B55-alpha|PP2A subunit B isoform PR55-alpha|PP2A subunit B isoform R2-alpha|PP2A subunit B PR55 alpha isoform|PP2A subunit B R2 alpha isoform|PPP2R2A|PR52A|PR55A|Protein phosphatase 2 (formerly 2A) regulatory subunit B (PR 52) alpha isoform|Protein phosphatase 2 (formerly 2A)|regulatory subunit B (PR52)|alpha isoform|Protein phosphatase 2 regulatory subunit B alpha|Protein phosphatase 2 regulatory subunit B alpha isoform|Protein phosphatase 2 regulatory subunit Balpha|Serine/threonine protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform|Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform|Testicular tissue protein Li 156
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P63151
- P63151
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together