Anti-PTAFR Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PTAFR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77795-951
- 77795-950
- Antigen symbol:
- PTAFR
- PTAFR
- Antigen name:
- Platelet-activating factor receptor protein (297-342AA)
- Platelet-activating factor receptor protein (297-342AA)
- Antigen synonyms:
- OTTHUMP00000004041| OTTMUSP00000010350| PAF R| PAF receptor| PAF-R| PAFr| Platelet activating factor receptor| Platelet-activating factor receptor| PTAFR| PTAFR_HUMAN| RP23-470B20.1
- OTTHUMP00000004041| OTTMUSP00000010350| PAF R| PAF receptor| PAF-R| PAFr| Platelet activating factor receptor| Platelet-activating factor receptor| PTAFR| PTAFR_HUMAN| RP23-470B20.1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25105
- P25105
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together