Anti-PSMD2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PSMD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-860
- 77823-859
- Antigen symbol:
- PSMD2
- PSMD2
- Antigen name:
- 26S proteasome non-ATPase regulatory subunit 2 protein (401-700AA)
- 26S proteasome non-ATPase regulatory subunit 2 protein (401-700AA)
- Antigen synonyms:
- 26S proteasome non-ATPase regulatory subunit 2|26S proteasome regulatory subunit RPN1|26S proteasome regulatory subunit S2|26S proteasome subunit p97|55.11 protein|MGC14274|P97|Proteasome (prosome macropain) 26S subunit non ATPase 2|Proteasome 26S subunit|non-ATPase|2|Proteasome 26S|subunit 2|Protein 55.11|Psmd2|PSMD2_HUMAN|Rpn1|S2|TNFR associated protein 2|TRAP2|Tumor necrosis factor receptor associated protein 2|Tumor necrosis factor type 1 receptor-associated protein 2
- 26S proteasome non-ATPase regulatory subunit 2|26S proteasome regulatory subunit RPN1|26S proteasome regulatory subunit S2|26S proteasome subunit p97|55.11 protein|MGC14274|P97|Proteasome (prosome macropain) 26S subunit non ATPase 2|Proteasome 26S subunit|non-ATPase|2|Proteasome 26S|subunit 2|Protein 55.11|Psmd2|PSMD2_HUMAN|Rpn1|S2|TNFR associated protein 2|TRAP2|Tumor necrosis factor receptor associated protein 2|Tumor necrosis factor type 1 receptor-associated protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13200
- Q13200
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together