Anti-ECHDC1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: ECHDC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77840-991
- 77840-992
- Antigen symbol:
- ECHDC1
- ECHDC1
- Antigen name:
- Ethylmalonyl-CoA decarboxylase protein (1-301AA)
- Ethylmalonyl-CoA decarboxylase protein (1-301AA)
- Antigen synonyms:
- ECHD1_HUMAN|ECHDC1|Enoyl CoA hydratase domain containing 1|Enoyl CoA hydratase domain containing protein 1|Enoyl Coenzyme A hydratase domain containing 1|Enoyl-CoA hydratase domain-containing protein 1|Uncharacterized hypothalamus protein HCDASE
- ECHD1_HUMAN|ECHDC1|Enoyl CoA hydratase domain containing 1|Enoyl CoA hydratase domain containing protein 1|Enoyl Coenzyme A hydratase domain containing 1|Enoyl-CoA hydratase domain-containing protein 1|Uncharacterized hypothalamus protein HCDASE
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NTX5
- Q9NTX5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together