Anti-TMSB4X Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TMSB4X
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-182
- 77803-182
- Antigen symbol:
- TMSB4X
- TMSB4X
- Antigen name:
- Thymosin beta-4 protein (6-44AA)
- Thymosin beta-4 protein (6-44AA)
- Antigen synonyms:
- Fx|Hematopoietic system regulatory peptide|Prothymosin beta 4|PTMB 4|PTMB4|Seraspenide|T beta 4|T beta-4|TB4X|THYB 4|Thyb4|Thymosin beta 4|Thymosin beta 4 X chromosome|Thymosin beta 4 X linked|TMSB 4|TMSB4|TYB4_HUMAN
- Fx|Hematopoietic system regulatory peptide|Prothymosin beta 4|PTMB 4|PTMB4|Seraspenide|T beta 4|T beta-4|TB4X|THYB 4|Thyb4|Thymosin beta 4|Thymosin beta 4 X chromosome|Thymosin beta 4 X linked|TMSB 4|TMSB4|TYB4_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P62328
- P62328
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together