Anti-TAF2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TAF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-027
- 77830-026
- Antigen symbol:
- TAF2
- TAF2
- Antigen name:
- Transcription initiation factor TFIID subunit 2 protein (400-450AA)
- Transcription initiation factor TFIID subunit 2 protein (400-450AA)
- Antigen synonyms:
- TAF2|CIF150|TAF2B|Transcription initiation factor TFIID subunit 2|150 kDa cofactor of initiator function|RNA polymerase II TBP-associated factor subunit B|TBP-associated factor 150 kDa|Transcription initiation factor TFIID 150 kDa subunit|TAF(II)150|TAFII-150|TAFII150
- TAF2|CIF150|TAF2B|Transcription initiation factor TFIID subunit 2|150 kDa cofactor of initiator function|RNA polymerase II TBP-associated factor subunit B|TBP-associated factor 150 kDa|Transcription initiation factor TFIID 150 kDa subunit|TAF(II)150|TAFII-150|TAFII150
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6P1X5
- Q6P1X5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together