Anti-ARPIN Rabbit Recombinant Polyclonal Antibody
Catalogue # 77832-605
Fournisseur: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Antigen name:Arpin protein (1-226AA)
- Antigen symbol:ARPIN
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q7Z6K5
- Antigen synonyms:2610034B18Rik|Actin related protein 2/3 complex inhibitor|Arp2/3 inhibition protein|ARPIN|C15orf38|Chromosome 15 open reading frame 38|CO038_HUMAN|Hypothetical protein LOC348110|MGC61550|RIKEN cDNA 2610034B18|UPF0552 protein C15orf38
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ARPIN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human