Anti-ERGIC2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ERGIC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-348
- 77834-347
- Antigen symbol:
- ERGIC2
- ERGIC2
- Antigen name:
- Endoplasmic reticulum-Golgi intermediate compartment protein 2 protein (55-310AA)
- Endoplasmic reticulum-Golgi intermediate compartment protein 2 protein (55-310AA)
- Antigen synonyms:
- cd002|CD14 protein|CDA14|Endoplasmic reticulum Golgi intermediate compartment protein 2|Endoplasmic reticulum-Golgi intermediate compartment protein 2|ERGI2_HUMAN|ERGIC 2|ERGIC and golgi 2|Erv41|MGC111152|PTX1|PTX1 protein
- cd002|CD14 protein|CDA14|Endoplasmic reticulum Golgi intermediate compartment protein 2|Endoplasmic reticulum-Golgi intermediate compartment protein 2|ERGI2_HUMAN|ERGIC 2|ERGIC and golgi 2|Erv41|MGC111152|PTX1|PTX1 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96RQ1
- Q96RQ1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together