Order Entry
Canada
ContactUsLinkComponent
 
Anti-NSDHL Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
 
undefined
Anti-NSDHL Rabbit Recombinant Polyclonal Antibody (HRP)
New Product

 

  • Catalog No:
  • 77823-694
  • 77823-693
  • Antigen symbol:
  • NSDHL
  • NSDHL
  • Antigen name:
  • Sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating protein (154-211AA)
  • Sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating protein (154-211AA)
  • Antigen synonyms:
  • decarboxylating|H105E3|H105e3 protein|NAD(P) dependent steroid dehydrogenase like|NSDHL|NSDHL_HUMAN|Protein H105e3|SDR31E1|Short chain dehydrogenase/reductase family 31E member 1|Sterol 4 alpha carboxylate 3 dehydrogenase decarboxylating|Sterol-4-alpha-carboxylate 3-dehydrogenase|XAP104
  • decarboxylating|H105E3|H105e3 protein|NAD(P) dependent steroid dehydrogenase like|NSDHL|NSDHL_HUMAN|Protein H105e3|SDR31E1|Short chain dehydrogenase/reductase family 31E member 1|Sterol 4 alpha carboxylate 3 dehydrogenase decarboxylating|Sterol-4-alpha-carboxylate 3-dehydrogenase|XAP104
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q15738
  • Q15738
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together