Anti-NPL Rabbit Recombinant Polyclonal Antibody (Biotin)
New Product
Type:
Antigen: NPL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77839-190
- 77839-189
- Antigen symbol:
- NPL
- NPL
- Antigen name:
- N-acetylneuraminate lyase protein (17-77AA)
- N-acetylneuraminate lyase protein (17-77AA)
- Antigen synonyms:
- 0610033B02Rik|C112|C1orf13|Dihydrodipicolinate synthase|Dihydrodipicolinate synthetase homolog 1|MGC149582|MGC61869|N-acetylneuraminate lyase|N-acetylneuraminate pyruvate-lyase|N-acetylneuraminic acid aldolase|NAL|NALase|NPL|NPL_HUMAN|NPL1|RP11-249O6.1|Sialate lyase|Sialate-pyruvate lyase|Sialic acid aldolase|Sialic acid lyase
- 0610033B02Rik|C112|C1orf13|Dihydrodipicolinate synthase|Dihydrodipicolinate synthetase homolog 1|MGC149582|MGC61869|N-acetylneuraminate lyase|N-acetylneuraminate pyruvate-lyase|N-acetylneuraminic acid aldolase|NAL|NALase|NPL|NPL_HUMAN|NPL1|RP11-249O6.1|Sialate lyase|Sialate-pyruvate lyase|Sialic acid aldolase|Sialic acid lyase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BXD5
- Q9BXD5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together