Anti-KRT73 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KRT73
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-767
- 77832-766
- Antigen symbol:
- KRT73
- KRT73
- Antigen name:
- Keratin, type II cytoskeletal 73 protein (1-300AA)
- Keratin, type II cytoskeletal 73 protein (1-300AA)
- Antigen synonyms:
- CK-73|Cytokeratin-73|IRT6IRS3|K2C73_HUMAN|K6IRS3|K73|Keratin 6 irs3|Keratin, type II cytoskeletal 73|Keratin-73|KRT6IRS3|Krt73|Type II inner root sheath-specific keratin-K6irs3|Type-II keratin Kb36
- CK-73|Cytokeratin-73|IRT6IRS3|K2C73_HUMAN|K6IRS3|K73|Keratin 6 irs3|Keratin, type II cytoskeletal 73|Keratin-73|KRT6IRS3|Krt73|Type II inner root sheath-specific keratin-K6irs3|Type-II keratin Kb36
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86Y46
- Q86Y46
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together