Anti-PQLC3 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: PQLC3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77833-390
- 77833-389
- Antigen symbol:
- PQLC3
- PQLC3
- Antigen name:
- PQ-loop repeat-containing protein 3 protein (118-170AA)
- PQ-loop repeat-containing protein 3 protein (118-170AA)
- Antigen synonyms:
- PQ-loop repeat-containing protein 3|PQLC3_HUMAN
- PQ-loop repeat-containing protein 3|PQLC3_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N755
- Q8N755
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together