Anti-MAPK7 Rabbit Recombinant Polyclonal Antibody
Catalog # 77812-541
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Recombinant Protein
- Antigen symbol:MAPK7
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q13164
- Antigen synonyms:Big MAP kinase 1|BMK 1|BMK 1 kinase|BMK-1|BMK1|BMK1 Kinase|EC 2.7.11.24|ERK 4|ERK 5|ERK-5|ERK4|ERK5|Extracellular signal regulated kinase 5|Extracellular signal-regulated kinase 5|MAP kinase 7|MAPK 7|Mitogen activated protein kinase 7|Mitogen-activated protein kinase 7|MK07_HUMAN|OTTHUMP00000065906|OTTHUMP00000065907|PRKM 7|PRKM7|PROTEIN KINASE|MITOGEN-ACTIVATED|7
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 2000, IHC: 1:200 to 500
Type:
Antigen: MAPK7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat