Order Entry
Canada
ContactUsLinkComponent
Anti-SLC6A16 Rabbit Recombinant Polyclonal Antibody
Anti-SLC6A16 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-SLC6A16 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77836-018
  • 77836-017
  • Antigen symbol:
  • SLC6A16
  • SLC6A16
  • Antigen name:
  • Orphan sodium- and chloride-dependent neurotransmitter transporter NTT5 protein (1-138AA)
  • Orphan sodium- and chloride-dependent neurotransmitter transporter NTT5 protein (1-138AA)
  • Antigen synonyms:
  • NT5|NTT5|Orphan sodium- and chloride-dependent neurotransmitter transporter NTT5|S6A16_HUMAN|SLC6A16|solute carrier family 6 (neurotransmitter transporter), member 16|Solute carrier family 6 member 16
  • NT5|NTT5|Orphan sodium- and chloride-dependent neurotransmitter transporter NTT5|S6A16_HUMAN|SLC6A16|solute carrier family 6 (neurotransmitter transporter), member 16|Solute carrier family 6 member 16
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9GZN6
  • Q9GZN6
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together