Anti-TNKS Rabbit Recombinant Antibody
Catalog # 77803-262
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Tankyrase-1 protein (35-130AA)
- Antigen symbol:TNKS
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O95271
- Antigen synonyms:ADP-ribosyltransferase diphtheria toxin-like 5|OTTHUMP00000115550|OTTHUMP00000224675|PARP 5a|PARP5A|PARPL|pART5|Poly [ADP-ribose] polymerase 5A|TANK 1|TANK1|Tankyrase 1|Tankyrase I|Tankyrase TRF1 interacting ankyrin related ADP ribose polymerase|Tankyrase-1|Tankyrase1|TankyraseI|TIN 1|TIN1|TINF 1|TINF1|TNKS 1|TNKS|TNKS-1|TNKS1|TNKS1_HUMAN|TRF1 interacting ankyrin related ADP ribose polymerase|TRF1-interacting ankyrin-related ADP-ribose polymerase
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: TNKS
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human