Anti-MRPL49 Rabbit Polyclonal Antibody
77809-147
New Product
- Antigen name:Synthesized peptide derived from the Internal region of MRP-L49
- Antigen symbol:MRPL49
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q13405
- Antigen synonyms:MRPL49|C11orf4|NOF1|OK/SW-cl.67|39S ribosomal protein L49|mitochondrial|L49mt|MRP-L49|Mitochondrial large ribosomal subunit protein mL49|Neighbor of FAU|NOF|Protein NOF1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: MRPL49
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human