Anti-CAND1 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: CAND1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77832-731
- 77832-730
- Antigen symbol:
- CAND1
- CAND1
- Antigen name:
- Cullin-associated NEDD8-dissociated protein 1 protein (1150-1230AA)
- Cullin-associated NEDD8-dissociated protein 1 protein (1150-1230AA)
- Antigen synonyms:
- CAND 1|CAND1_HUMAN|Cullin associated and neddylation dissociated 1|Cullin associated and neddylation dissociated protein 1|Cullin associated NEDD8 dissociated protein 1|Cullin-associated and neddylation-dissociated protein 1|Cullin-associated NEDD8-dissociated protein 1|DKFZP434M1414|FLJ10114|FLJ10929|FLJ38691|FLJ90441|KIAA0829|p120 TATA binding protein interacting protein 120A|TBP interacting protein|TBP interacting protein of 120 kDa A|TBP Interacting Protein TIP120A|TBP-interacting protein 120A|TBP-interacting protein of 120 kDa A|TIP120|TIP120 protein|TIP120A
- CAND 1|CAND1_HUMAN|Cullin associated and neddylation dissociated 1|Cullin associated and neddylation dissociated protein 1|Cullin associated NEDD8 dissociated protein 1|Cullin-associated and neddylation-dissociated protein 1|Cullin-associated NEDD8-dissociated protein 1|DKFZP434M1414|FLJ10114|FLJ10929|FLJ38691|FLJ90441|KIAA0829|p120 TATA binding protein interacting protein 120A|TBP interacting protein|TBP interacting protein of 120 kDa A|TBP Interacting Protein TIP120A|TBP-interacting protein 120A|TBP-interacting protein of 120 kDa A|TIP120|TIP120 protein|TIP120A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86VP6
- Q86VP6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together