Anti-GNPDA2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: GNPDA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77833-578
- 77833-579
- Antigen symbol:
- GNPDA2
- GNPDA2
- Antigen name:
- Glucosamine-6-phosphate isomerase 2 protein (28-154AA)
- Glucosamine-6-phosphate isomerase 2 protein (28-154AA)
- Antigen synonyms:
- GlcN6P deaminase 2|Glucosamine 6 phosphate deaminase 2|Glucosamine 6 phosphate isomerase 2|Glucosamine 6 phosphate isomerase SB52|Glucosamine-6-phosphate deaminase 2|Glucosamine-6-phosphate isomerase 2|Glucosamine-6-phosphate isomerase SB52|GNP2|GNPDA 2|Gnpda2|GNPI2_HUMAN|SB52
- GlcN6P deaminase 2|Glucosamine 6 phosphate deaminase 2|Glucosamine 6 phosphate isomerase 2|Glucosamine 6 phosphate isomerase SB52|Glucosamine-6-phosphate deaminase 2|Glucosamine-6-phosphate isomerase 2|Glucosamine-6-phosphate isomerase SB52|GNP2|GNPDA 2|Gnpda2|GNPI2_HUMAN|SB52
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TDQ7
- Q8TDQ7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together