Order Entry
Canada
ContactUsLinkComponent
Anti-FOXD4 Rabbit Polyclonal Antibody
Anti-FOXD4 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-FOXD4 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77821-153
  • 77821-152
  • Antigen symbol:
  • FOXD4
  • FOXD4
  • Antigen name:
  • Synthetic peptide of FOXD4
  • Synthetic peptide of FOXD4
  • Antigen synonyms:
  • FKHL9|Forkhead box D4|Forkhead box protein D4|Forkhead Drosophila homolog like 9|Forkhead like 9|Forkhead related activator 5|Forkhead related protein FKHL9|Forkhead related transcription factor 5|Forkhead-related protein FKHL9|Forkhead-related transcription factor 5|FOXD4|FOXD4_HUMAN|FOXD4A|FREAC 5|FREAC-5|FREAC5|MGC105106|Myeloid factor alpha|Myeloid factor-alpha
  • FKHL9|Forkhead box D4|Forkhead box protein D4|Forkhead Drosophila homolog like 9|Forkhead like 9|Forkhead related activator 5|Forkhead related protein FKHL9|Forkhead related transcription factor 5|Forkhead-related protein FKHL9|Forkhead-related transcription factor 5|FOXD4|FOXD4_HUMAN|FOXD4A|FREAC 5|FREAC-5|FREAC5|MGC105106|Myeloid factor alpha|Myeloid factor-alpha
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q12950
  • Q12950
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together