Anti-GRIN2C Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:10000, IHC: 1:100 to 1:300
Type:
Antigen: GRIN2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77820-954
- 77820-953
- Antigen symbol:
- GRIN2C
- GRIN2C
- Antigen name:
- Synthetic peptide of GRIN2C
- Synthetic peptide of GRIN2C
- Antigen synonyms:
- GluN2C|Glutamate [NMDA] receptor subunit epsilon-3|Glutamate Receptor Ionotropic N Methyl D Aspartate 2C|Glutamate receptor ionotropic, NMDA 2C|Glutamate receptor, ionotropic, N methyl D aspartate 2C|Grin2c|N Methly D Aspartate Receptor Channel Subunit Epsilon 3|N Methyl D Aspartate Receptor Subtype 2C|N methyl D aspartate receptor subunit 2C|N-methyl D-aspartate receptor subtype 2C|NMDAR2C|NMDE3_HUMAN|NR2C|OTTHUMP00000041930
- GluN2C|Glutamate [NMDA] receptor subunit epsilon-3|Glutamate Receptor Ionotropic N Methyl D Aspartate 2C|Glutamate receptor ionotropic, NMDA 2C|Glutamate receptor, ionotropic, N methyl D aspartate 2C|Grin2c|N Methly D Aspartate Receptor Channel Subunit Epsilon 3|N Methyl D Aspartate Receptor Subtype 2C|N methyl D aspartate receptor subunit 2C|N-methyl D-aspartate receptor subtype 2C|NMDAR2C|NMDE3_HUMAN|NR2C|OTTHUMP00000041930
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14957
- Q14957
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together