Anti-LAPTM5 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LAPTM5
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-777
- 77824-776
- Antigen symbol:
- LAPTM5
- LAPTM5
- Antigen name:
- Lysosomal-associated transmembrane protein 5 protein (205-262AA)
- Lysosomal-associated transmembrane protein 5 protein (205-262AA)
- Antigen synonyms:
- LAPTM5|KIAA0085|Lysosomal-associated transmembrane protein 5|Lysosomal-associated multitransmembrane protein 5|Retinoic acid-inducible E3 protein
- LAPTM5|KIAA0085|Lysosomal-associated transmembrane protein 5|Lysosomal-associated multitransmembrane protein 5|Retinoic acid-inducible E3 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13571
- Q13571
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together