Anti-BPIFB2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:10000, WB:1:1000 to 1:5000, IHC:1:25 to 1:100
Type:
Antigen: BPIFB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77806-573
- 77806-572
- Antigen symbol:
- BPIFB2
- BPIFB2
- Antigen name:
- Synthetic peptide of BPIFB2
- Synthetic peptide of BPIFB2
- Antigen synonyms:
- Bactericidal permeability increasing protein like 1|Bactericidal/permeability-increasing protein-like 1|BPI fold-containing family B member 2|BPI-like 1|BPIB2_HUMAN|Bpifb2|C20orf184|dJ726C3.2|Long palate|Long palate lung and nasal epithelium carcinoma associated 2|Long palate lung and nasal epithelium carcinoma associated protein 2|LPLUNC2|lung and nasal epithelium carcinoma-associated protein 2|OTTHUMP00000030612|RYSR
- Bactericidal permeability increasing protein like 1|Bactericidal/permeability-increasing protein-like 1|BPI fold-containing family B member 2|BPI-like 1|BPIB2_HUMAN|Bpifb2|C20orf184|dJ726C3.2|Long palate|Long palate lung and nasal epithelium carcinoma associated 2|Long palate lung and nasal epithelium carcinoma associated protein 2|LPLUNC2|lung and nasal epithelium carcinoma-associated protein 2|OTTHUMP00000030612|RYSR
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N4F0
- Q8N4F0
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together