Anti-DHX30 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DHX30
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-477
- 77830-478
- Antigen symbol:
- DHX30
- DHX30
- Antigen name:
- Putative ATP-dependent RNA helicase DHX30 protein (694-1000AA)
- Putative ATP-dependent RNA helicase DHX30 protein (694-1000AA)
- Antigen synonyms:
- DDX30|DEAH box protein 30|Dhx30|DHX30_HUMAN|FLJ11214|KIAA0890 |Putative ATP-dependent RNA helicase DHX30|Ret CoR
- DDX30|DEAH box protein 30|Dhx30|DHX30_HUMAN|FLJ11214|KIAA0890 |Putative ATP-dependent RNA helicase DHX30|Ret CoR
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7L2E3
- Q7L2E3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together