Anti-STK38 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: STK38
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77822-625
- 77822-624
- Antigen symbol:
- STK38
- STK38
- Antigen name:
- Serine/threonine-protein kinase 38 protein (1-200AA)
- Serine/threonine-protein kinase 38 protein (1-200AA)
- Antigen synonyms:
- NDR 1|NDR|Ndr Ser/Thr kinase-like protein|NDR1|NDR1 protein kinase|Nuclear Dbf2 related kinase 1|nuclear Dbf2-related 1|Nuclear Dbf2-related kinase 1|Nuclear Dbf2-related protein|Serine/threonine protein kinase 38|Serine/threonine-protein kinase 38|STK 38|STK38|STK38_HUMAN
- NDR 1|NDR|Ndr Ser/Thr kinase-like protein|NDR1|NDR1 protein kinase|Nuclear Dbf2 related kinase 1|nuclear Dbf2-related 1|Nuclear Dbf2-related kinase 1|Nuclear Dbf2-related protein|Serine/threonine protein kinase 38|Serine/threonine-protein kinase 38|STK 38|STK38|STK38_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15208
- Q15208
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together