Order Entry
Canada
ContactUsLinkComponent
Anti-ST3GAL4 Rabbit Recombinant Polyclonal Antibody (HRP)
Anti-ST3GAL4 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-ST3GAL4 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77821-740
  • 77821-741
  • Antigen symbol:
  • ST3GAL4
  • ST3GAL4
  • Antigen name:
  • CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4 protein (39-184AA)
  • CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4 protein (39-184AA)
  • Antigen synonyms:
  • CGS23|NANTA3|SIAT4C|STZ|CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4|Alpha 2,3-ST 4|Beta-galactoside alpha-2,3-sialyltransferase 4|EC 2.4.99.2|EC 2.4.99.4|Alpha 2,3-sialyltransferase IV|Gal-NAc6S|Gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase|SAT-3|ST-4|ST3Gal IV|ST3GalIV|ST3GalA.2|Sialyltransferase 4C|SIAT4-C
  • CGS23|NANTA3|SIAT4C|STZ|CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 4|Alpha 2,3-ST 4|Beta-galactoside alpha-2,3-sialyltransferase 4|EC 2.4.99.2|EC 2.4.99.4|Alpha 2,3-sialyltransferase IV|Gal-NAc6S|Gal-beta-1,4-GalNAc-alpha-2,3-sialyltransferase|SAT-3|ST-4|ST3Gal IV|ST3GalIV|ST3GalA.2|Sialyltransferase 4C|SIAT4-C
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q11206
  • Q11206
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together