Order Entry
Canada
ContactUsLinkComponent
Anti-TAF5L Rabbit Recombinant Antibody
Anti-TAF5L Rabbit Recombinant Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-TAF5L Rabbit Recombinant Antibody
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77801-807
  • 77801-808
  • Antigen symbol:
  • TAF5L
  • TAF5L
  • Antigen name:
  • TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L protein (1-325AA)
  • TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L protein (1-325AA)
  • Antigen synonyms:
  • 1110005N04Rik|AI849020|OTTHUMP00000037470|OTTHUMP00000037471|PAF65 beta|PAF65-beta|PAF65B|PCAF associated factor 65 beta|PCAF-associated factor 65 beta|TAF 5L|TAF5 like RNA polymerase II p300/CBP associated factor (PCAF) associated factor 65 kD|TAF5 like RNA polymerase II p300/CBP associated factor (PCAF) associated factor 65 kDa|TAF5 like RNA polymerase II p300/CBP associated factor (PCAF) associated factor|TAF5 like RNA polymerase II p300/CBP associated factor associated factor 65 kDa subunit 5L|TAF5 like RNA polymerase II p300/CBP associated factor associated factor 65kDa|TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L|Taf5l|Taf5l protein|TAF5L_HUMAN
  • 1110005N04Rik|AI849020|OTTHUMP00000037470|OTTHUMP00000037471|PAF65 beta|PAF65-beta|PAF65B|PCAF associated factor 65 beta|PCAF-associated factor 65 beta|TAF 5L|TAF5 like RNA polymerase II p300/CBP associated factor (PCAF) associated factor 65 kD|TAF5 like RNA polymerase II p300/CBP associated factor (PCAF) associated factor 65 kDa|TAF5 like RNA polymerase II p300/CBP associated factor (PCAF) associated factor|TAF5 like RNA polymerase II p300/CBP associated factor associated factor 65 kDa subunit 5L|TAF5 like RNA polymerase II p300/CBP associated factor associated factor 65kDa|TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L|Taf5l|Taf5l protein|TAF5L_HUMAN
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O75529
  • O75529
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage buffer:
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together