Anti-DTX1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: DTX1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-437
- 77832-436
- Antigen symbol:
- DTX1
- DTX1
- Antigen name:
- E3 ubiquitin-protein ligase DTX1 protein (261-410AA)
- E3 ubiquitin-protein ligase DTX1 protein (261-410AA)
- Antigen synonyms:
- Deltex 1|E3 ubiquitin ligase|Deltex|Deltex homolog 1 (Drosophila)|Deltex homolog 1|Deltex protein 1|Deltex-1|Deltex1|dtx1|DTX1_HUMAN|E3 ubiquitin protein ligase DTX1|E3 ubiquitin-protein ligase DTX1|FXI-T1|Fxit 1|Fxit1|hDTX1|hDx 1|mDTX1|Protein deltex 1|Protein deltex-1
- Deltex 1|E3 ubiquitin ligase|Deltex|Deltex homolog 1 (Drosophila)|Deltex homolog 1|Deltex protein 1|Deltex-1|Deltex1|dtx1|DTX1_HUMAN|E3 ubiquitin protein ligase DTX1|E3 ubiquitin-protein ligase DTX1|FXI-T1|Fxit 1|Fxit1|hDTX1|hDx 1|mDTX1|Protein deltex 1|Protein deltex-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86Y01
- Q86Y01
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together