Anti-BCL11A Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: BCL11A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-275
- 77838-274
- Antigen symbol:
- BCL11A
- BCL11A
- Antigen name:
- B-cell lymphoma/leukemia 11A protein (1-230AA)
- B-cell lymphoma/leukemia 11A protein (1-230AA)
- Antigen synonyms:
- 2810047E18Rik|B cell CLL/lymphoma 11A (zinc finger protein)|B cell CLL/lymphoma 11A (zinc finger protein) isoform 2|B-cell CLL/lymphoma 11A|B-cell lymphoma/leukemia 11A|BC11A_HUMAN|BCL-11A|BCL11A B cell CLL/lymphoma 11A (zinc finger protein) isoform 1|BCL11A L|BCL11A S|BCL11A XL|BCL11a-M|BCL11AL|BCL11AS|BCL11AXL|C2H2 type zinc finger protein|COUP TF interacting protein 1|COUP-TF-interacting protein 1|CTIP1|mouse|homolog of|D930021L15Rik|Ecotropic viral integration site 9|Ecotropic viral integration site 9 homolog|Ecotropic viral integration site 9 protein|Ecotropic viral integration site 9 protein homolog|EVI-9|Evi9|FLJ10173|FLJ34997|HBFQTL5|KIAA1809|mKIAA1809|OTTHUMP00000159788|OTTHUMP00000159789|OTTHUMP00000201250|OTTHUMP00000202084|Zinc finger protein 856|ZNF856
- 2810047E18Rik|B cell CLL/lymphoma 11A (zinc finger protein)|B cell CLL/lymphoma 11A (zinc finger protein) isoform 2|B-cell CLL/lymphoma 11A|B-cell lymphoma/leukemia 11A|BC11A_HUMAN|BCL-11A|BCL11A B cell CLL/lymphoma 11A (zinc finger protein) isoform 1|BCL11A L|BCL11A S|BCL11A XL|BCL11a-M|BCL11AL|BCL11AS|BCL11AXL|C2H2 type zinc finger protein|COUP TF interacting protein 1|COUP-TF-interacting protein 1|CTIP1|mouse|homolog of|D930021L15Rik|Ecotropic viral integration site 9|Ecotropic viral integration site 9 homolog|Ecotropic viral integration site 9 protein|Ecotropic viral integration site 9 protein homolog|EVI-9|Evi9|FLJ10173|FLJ34997|HBFQTL5|KIAA1809|mKIAA1809|OTTHUMP00000159788|OTTHUMP00000159789|OTTHUMP00000201250|OTTHUMP00000202084|Zinc finger protein 856|ZNF856
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H165
- Q9H165
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together