Anti-RAF1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-588
- 77796-587
- Antigen symbol:
- RAF1
- RAF1
- Antigen name:
- RAF proto-oncogene serine/threonine-protein kinase protein (1-648AA)
- RAF proto-oncogene serine/threonine-protein kinase protein (1-648AA)
- Antigen synonyms:
- c Raf|C-raf|C-Raf proto-oncogene, serine/threonine kinase|CMD1NN|Craf 1 transforming gene|cRaf|Craf1 transforming gene|EC 2.7.11.1|kinase Raf1|Murine sarcoma 3611 oncogene 1|NS5|Oncogene MIL|Oncogene RAF1|OTTHUMP00000160218|OTTHUMP00000207813|OTTHUMP00000209389|Protein kinase raf 1|Proto-oncogene c-RAF|Raf 1|Raf 1 proto oncogene serine/threonine kinase|RAF|Raf proto oncogene serine/threonine protein kinase|RAF proto-oncogene serine/threonine-protein kinase|RAF-1|RAF1|RAF1_HUMAN|Similar to murine leukemia viral (V-raf-1) oncogene homolog 1|TRANSFORMING REPLICATION-DEFECTIVE MURINE RETROVIRUS 3611-MSV|v raf 1 murine leukemia viral oncogene homolog 1|v-raf murine sarcoma viral oncogene homolog 1|v-raf-1 murine leukemia viral oncogene-like protein 1|vraf1 murine leukemia viral oncogene homolog 1
- c Raf|C-raf|C-Raf proto-oncogene, serine/threonine kinase|CMD1NN|Craf 1 transforming gene|cRaf|Craf1 transforming gene|EC 2.7.11.1|kinase Raf1|Murine sarcoma 3611 oncogene 1|NS5|Oncogene MIL|Oncogene RAF1|OTTHUMP00000160218|OTTHUMP00000207813|OTTHUMP00000209389|Protein kinase raf 1|Proto-oncogene c-RAF|Raf 1|Raf 1 proto oncogene serine/threonine kinase|RAF|Raf proto oncogene serine/threonine protein kinase|RAF proto-oncogene serine/threonine-protein kinase|RAF-1|RAF1|RAF1_HUMAN|Similar to murine leukemia viral (V-raf-1) oncogene homolog 1|TRANSFORMING REPLICATION-DEFECTIVE MURINE RETROVIRUS 3611-MSV|v raf 1 murine leukemia viral oncogene homolog 1|v-raf murine sarcoma viral oncogene homolog 1|v-raf-1 murine leukemia viral oncogene-like protein 1|vraf1 murine leukemia viral oncogene homolog 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P04049
- P04049
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together