Anti-LTV1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LTV1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-027
- 77833-028
- Antigen symbol:
- LTV1
- LTV1
- Antigen name:
- Protein LTV1 homolog protein (201-475AA)
- Protein LTV1 homolog protein (201-475AA)
- Antigen synonyms:
- C6orf93|Chromosome 6 open reading frame 93|dJ468K18.4|FLJ14909|FLJ18519|Ltv1|LTV1 homolog (S. cerevisiae)|LTV1_HUMAN|OTTHUMP00000017340|Protein LTV1 homolog
- C6orf93|Chromosome 6 open reading frame 93|dJ468K18.4|FLJ14909|FLJ18519|Ltv1|LTV1 homolog (S. cerevisiae)|LTV1_HUMAN|OTTHUMP00000017340|Protein LTV1 homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96GA3
- Q96GA3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together