Anti-PFKM Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PFKM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77793-875
- 77793-876
- Antigen symbol:
- PFKM
- PFKM
- Antigen name:
- ATP-dependent 6-phosphofructokinase, muscle type protein (166-334AA)
- ATP-dependent 6-phosphofructokinase, muscle type protein (166-334AA)
- Antigen synonyms:
- 6 Phosphofructokinase Muscle Type|6-phosphofructokinase|6-phosphofructokinase muscle type|EC 2.7.1.1|EC 2.7.1.11|GSD7|K6PF_HUMAN|MGC8699|muscle type|PFK|PFK-A|PFKA|PFKL|PFKM|PFKP|PFKX|Phosphofructo 1 Kinase Isozyme A|Phosphofructo-1-kinase isozyme A|Phosphofructokinase 1|Phosphofructokinase M|Phosphofructokinase|muscle|polypeptide X|Phosphofructokinase-M|Phosphohexokinase
- 6 Phosphofructokinase Muscle Type|6-phosphofructokinase|6-phosphofructokinase muscle type|EC 2.7.1.1|EC 2.7.1.11|GSD7|K6PF_HUMAN|MGC8699|muscle type|PFK|PFK-A|PFKA|PFKL|PFKM|PFKP|PFKX|Phosphofructo 1 Kinase Isozyme A|Phosphofructo-1-kinase isozyme A|Phosphofructokinase 1|Phosphofructokinase M|Phosphofructokinase|muscle|polypeptide X|Phosphofructokinase-M|Phosphohexokinase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08237
- P08237
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together