Anti-SNRPD2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: SNRPD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77804-907
- 77804-906
- Antigen symbol:
- SNRPD2
- SNRPD2
- Antigen name:
- Small nuclear ribonucleoprotein Sm D2 protein (1-118AA)
- Small nuclear ribonucleoprotein Sm D2 protein (1-118AA)
- Antigen synonyms:
- Sm-D2|small nuclear ribonucleoprotein D2|Small nuclear ribonucleoprotein D2 (RCG54604)|small nuclear ribonucleoprotein D2 polypeptide (16.5kD)|small nuclear ribonucleoprotein D2 polypeptide 16.5kDa|SMALL NUCLEAR RIBONUCLEOPROTEIN POLYPEPTIDE D2, SNRPD2|Small nuclear ribonucleoprotein Sm D2|SMD2|SMD2_HUMAN|snRNP core protein D2|SNRPD1|SNRPD2 small nuclear ribonucleoprotein D2 polypeptide 16.5kDa
- Sm-D2|small nuclear ribonucleoprotein D2|Small nuclear ribonucleoprotein D2 (RCG54604)|small nuclear ribonucleoprotein D2 polypeptide (16.5kD)|small nuclear ribonucleoprotein D2 polypeptide 16.5kDa|SMALL NUCLEAR RIBONUCLEOPROTEIN POLYPEPTIDE D2, SNRPD2|Small nuclear ribonucleoprotein Sm D2|SMD2|SMD2_HUMAN|snRNP core protein D2|SNRPD1|SNRPD2 small nuclear ribonucleoprotein D2 polypeptide 16.5kDa
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P62316
- P62316
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together