Anti-SLC10A2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC10A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-689
- 77824-688
- Antigen symbol:
- SLC10A2
- SLC10A2
- Antigen name:
- Ileal sodium/bile acid cotransporter protein (306-348AA)
- Ileal sodium/bile acid cotransporter protein (306-348AA)
- Antigen synonyms:
- Apical sodium dependent bile acid transporter|Apical sodium- dependent bile acid transporter|Apical sodium-dependent bile acid transporter|ASBT|IBAT|ileal|ileal apical sodium-dependent bile acid transporter|Ileal Na(+)/bile acid cotransporter|Ileal sodium dependent bile acid transporter|Ileal sodium-dependent bile acid transporter|Ileal sodium/bile acid cotransporter|ISBT|Na(+)-dependent ileal bile acid transporter|Na+ bile acid cotransporter|Na+ dependent ileal bile acid transporter|NTCP2|NTCP2_HUMAN|PBAM|SLC10A2|Sodium/taurocholate cotransporting polypeptide|solute carrier family 10 (sodium/bile acid cotransporter family)|Solute carrier family 10 (sodium/bile acid cotransporter family)|member 2|Solute carrier family 10 member 2
- Apical sodium dependent bile acid transporter|Apical sodium- dependent bile acid transporter|Apical sodium-dependent bile acid transporter|ASBT|IBAT|ileal|ileal apical sodium-dependent bile acid transporter|Ileal Na(+)/bile acid cotransporter|Ileal sodium dependent bile acid transporter|Ileal sodium-dependent bile acid transporter|Ileal sodium/bile acid cotransporter|ISBT|Na(+)-dependent ileal bile acid transporter|Na+ bile acid cotransporter|Na+ dependent ileal bile acid transporter|NTCP2|NTCP2_HUMAN|PBAM|SLC10A2|Sodium/taurocholate cotransporting polypeptide|solute carrier family 10 (sodium/bile acid cotransporter family)|Solute carrier family 10 (sodium/bile acid cotransporter family)|member 2|Solute carrier family 10 member 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12908
- Q12908
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together