Anti-PHOX2A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PHOX2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77794-062
- 77794-063
- Antigen symbol:
- PHOX2A
- PHOX2A
- Antigen name:
- Paired mesoderm homeobox protein 2A protein (150-264AA)
- Paired mesoderm homeobox protein 2A protein (150-264AA)
- Antigen synonyms:
- Aristaless homeobox (Drosophila) fibrosis of extraocular muscles congenital 2 autosomal recessive|Aristaless homeobox gene homolog (Drosophila)|Aristaless homeobox homolog|Aristaless homeobox protein homolog|ARIX 1 homeodomain protein|ARIX|Arix homeodomain protein|ARIX1 homeodomain protein|CFEOM 2|CFEOM2|FEOM 2|FEOM2|Fibrosis of extraocular muscles congenital 2 autosomal recessive|MGC52227|NCAM 2|NCAM2|Paired like (aristaless) Homeobox 2A|Paired like homeobox 2a|Paired mesoderm homeobox 2a|Paired mesoderm homeobox protein 2A|Paired-like homeobox 2A|PHOX 2A|Phox2|Phox2a|PHX2A_HUMAN|Pmx 2a|Pmx2|Pmx2a
- Aristaless homeobox (Drosophila) fibrosis of extraocular muscles congenital 2 autosomal recessive|Aristaless homeobox gene homolog (Drosophila)|Aristaless homeobox homolog|Aristaless homeobox protein homolog|ARIX 1 homeodomain protein|ARIX|Arix homeodomain protein|ARIX1 homeodomain protein|CFEOM 2|CFEOM2|FEOM 2|FEOM2|Fibrosis of extraocular muscles congenital 2 autosomal recessive|MGC52227|NCAM 2|NCAM2|Paired like (aristaless) Homeobox 2A|Paired like homeobox 2a|Paired mesoderm homeobox 2a|Paired mesoderm homeobox protein 2A|Paired-like homeobox 2A|PHOX 2A|Phox2|Phox2a|PHX2A_HUMAN|Pmx 2a|Pmx2|Pmx2a
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14813
- O14813
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together