Anti-TFEC Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TFEC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77802-320
- 77802-319
- Antigen symbol:
- TFEC
- TFEC
- Antigen name:
- Transcription factor EC protein (1-197AA)
- Transcription factor EC protein (1-197AA)
- Antigen synonyms:
- bHLHe34|Class E basic helix-loop-helix protein 34|hTFEC-L|TCFEC|TFE-C|Tfec|TFEC protein|TFEC_HUMAN|TFECL|Transcription factor EC|Transcription factor EC-like
- bHLHe34|Class E basic helix-loop-helix protein 34|hTFEC-L|TCFEC|TFE-C|Tfec|TFEC protein|TFEC_HUMAN|TFECL|Transcription factor EC|Transcription factor EC-like
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14948
- O14948
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together