Anti-STRA6 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: STRA6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-871
- 77836-870
- Antigen symbol:
- STRA6
- STRA6
- Antigen name:
- Receptor for retinol uptake STRA6 protein (530-667AA)
- Receptor for retinol uptake STRA6 protein (530-667AA)
- Antigen synonyms:
- MCOPCB8|MCOPS9|PP14296|Stimulated by retinoic acid 6 homolog|Stimulated by retinoic acid gene 6 protein homolog|STRA6|STRA6_HUMAN
- MCOPCB8|MCOPS9|PP14296|Stimulated by retinoic acid 6 homolog|Stimulated by retinoic acid gene 6 protein homolog|STRA6|STRA6_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BX79
- Q9BX79
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together