Anti-PRPF39 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: PRPF39
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77831-930
- 77831-931
- Antigen symbol:
- PRPF39
- PRPF39
- Antigen name:
- Pre-mRNA-processing factor 39 protein (41-190AA)
- Pre-mRNA-processing factor 39 protein (41-190AA)
- Antigen synonyms:
- FLJ11128|FLJ20666|FLJ45460|MGC149842|MGC149843|Pre-mRNA-processing factor 39|PRP39 homolog|PRP39 pre-mRNA processing factor 39 homolog (S. cerevisiae)|PRP39_HUMAN|prpf39
- FLJ11128|FLJ20666|FLJ45460|MGC149842|MGC149843|Pre-mRNA-processing factor 39|PRP39 homolog|PRP39 pre-mRNA processing factor 39 homolog (S. cerevisiae)|PRP39_HUMAN|prpf39
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86UA1
- Q86UA1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together