Anti-PDCD10 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: PDCD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77818-836
- 77818-837
- Antigen symbol:
- PDCD10
- PDCD10
- Antigen name:
- Fusion protein of PDCD10
- Fusion protein of PDCD10
- Antigen synonyms:
- Apoptosis related protein 15|CCM3|Cerebral cavernous malformations 3 protein|MGC1212|MGC24477|PDC10_HUMAN|PDCD 10|PDCD10|Programmed cell death 10|Programmed cell death protein 10|TF 1 cell apoptosis related protein 15|TF-1 cell apoptosis-related protein 15|TFAR15
- Apoptosis related protein 15|CCM3|Cerebral cavernous malformations 3 protein|MGC1212|MGC24477|PDC10_HUMAN|PDCD 10|PDCD10|Programmed cell death 10|Programmed cell death protein 10|TF 1 cell apoptosis related protein 15|TF-1 cell apoptosis-related protein 15|TFAR15
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BUL8
- Q9BUL8
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together