Anti-TP53BP2 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:200 to 1:1000
Type:
Antigen: TP53BP2
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77824-788
- 77824-789
- Antigen symbol:
- TP53BP2
- TP53BP2
- Antigen name:
- Apoptosis-stimulating of p53 protein 2 protein (360-600AA)
- Apoptosis-stimulating of p53 protein 2 protein (360-600AA)
- Antigen synonyms:
- 53BP2|Apoptosis stimulating of p53 protein 2|Apoptosis stimulating protein of p53 2|Apoptosis-stimulating of p53 protein 2|ASPP2|ASPP2_HUMAN|BBP|Bcl2 binding protein|Bcl2-binding protein|NY REN 51 antigen|p53 binding protein 2|p53-binding protein 2|p53BP2|PPP1R13A|Renal carcinoma antigen NY-REN-51|Tp53bp2|Tumor protein p53 binding protein 2|Tumor suppressor p53 binding protein 2|Tumor suppressor p53-binding protein 2
- 53BP2|Apoptosis stimulating of p53 protein 2|Apoptosis stimulating protein of p53 2|Apoptosis-stimulating of p53 protein 2|ASPP2|ASPP2_HUMAN|BBP|Bcl2 binding protein|Bcl2-binding protein|NY REN 51 antigen|p53 binding protein 2|p53-binding protein 2|p53BP2|PPP1R13A|Renal carcinoma antigen NY-REN-51|Tp53bp2|Tumor protein p53 binding protein 2|Tumor suppressor p53 binding protein 2|Tumor suppressor p53-binding protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13625
- Q13625
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together