Anti-TXNDC12 Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TXNDC12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-750
- 77803-750
- Antigen symbol:
- TXNDC12
- TXNDC12
- Antigen name:
- Thioredoxin domain-containing protein 12 protein (27-172AA)
- Thioredoxin domain-containing protein 12 protein (27-172AA)
- Antigen synonyms:
- AG1|AGR1|anterior gradient homolog 1|endoplasmic reticulum protein ERp19|Endoplasmic reticulum resident protein 18|Endoplasmic reticulum resident protein 19|endoplasmic reticulum thioredoxin superfamily member, 18 kDa|ER protein 18|ER protein 19|ERP 18|ERP16|ERp18|ERp19|hAG 1|hAG1|hTLP19|PDIA16|protein disulfide isomerase family A, member 16|thioredoxin domain containing 12 (endoplasmic reticulum|Thioredoxin domain-containing protein 12|thioredoxin like protein p19|Thioredoxin-like protein p19|TLP19|TXD12_HUMAN|TXNDC12
- AG1|AGR1|anterior gradient homolog 1|endoplasmic reticulum protein ERp19|Endoplasmic reticulum resident protein 18|Endoplasmic reticulum resident protein 19|endoplasmic reticulum thioredoxin superfamily member, 18 kDa|ER protein 18|ER protein 19|ERP 18|ERP16|ERp18|ERp19|hAG 1|hAG1|hTLP19|PDIA16|protein disulfide isomerase family A, member 16|thioredoxin domain containing 12 (endoplasmic reticulum|Thioredoxin domain-containing protein 12|thioredoxin like protein p19|Thioredoxin-like protein p19|TLP19|TXD12_HUMAN|TXNDC12
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95881
- O95881
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together