Anti-MRPL49 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MRPL49
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77821-867
- 77821-866
- Antigen symbol:
- MRPL49
- MRPL49
- Antigen name:
- 39S ribosomal protein L49, mitochondrial protein (1-166AA)
- 39S ribosomal protein L49, mitochondrial protein (1-166AA)
- Antigen synonyms:
- MRPL49|C11orf4|NOF1|OK/SW-cl.67|39S ribosomal protein L49|mitochondrial|L49mt|MRP-L49|Mitochondrial large ribosomal subunit protein mL49|Neighbor of FAU|NOF|Protein NOF1
- MRPL49|C11orf4|NOF1|OK/SW-cl.67|39S ribosomal protein L49|mitochondrial|L49mt|MRP-L49|Mitochondrial large ribosomal subunit protein mL49|Neighbor of FAU|NOF|Protein NOF1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13405
- Q13405
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together