Anti-BAZ2B Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:500 to 1:1000, IF: 1:200 to 1:500
Type:
Antigen: BAZ2B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-499
- 77841-500
- Antigen symbol:
- BAZ2B
- BAZ2B
- Antigen name:
- Bromodomain adjacent to zinc finger domain protein 2B protein (139-240AA)
- Bromodomain adjacent to zinc finger domain protein 2B protein (139-240AA)
- Antigen synonyms:
- BAZ2B|BAZ2B_HUMAN|Bromodomain adjacent to zinc finger domain 2B|Bromodomain adjacent to zinc finger domain protein 2B|DKFZp434H071|DKFZp762I0516|FLJ45644|hWALp4|OTTHUMP00000162897|OTTHUMP00000162898|WALp4
- BAZ2B|BAZ2B_HUMAN|Bromodomain adjacent to zinc finger domain 2B|Bromodomain adjacent to zinc finger domain protein 2B|DKFZp434H071|DKFZp762I0516|FLJ45644|hWALp4|OTTHUMP00000162897|OTTHUMP00000162898|WALp4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UIF8
- Q9UIF8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together