Anti-CBFB Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CBFB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-261
- 77825-262
- Antigen symbol:
- CBFB
- CBFB
- Antigen name:
- Core-binding factor subunit beta protein (1-182AA)
- Core-binding factor subunit beta protein (1-182AA)
- Antigen synonyms:
- CBF b|CBF beta|CBF-beta|CBFB|CBFbeta|Core binding factor beta subunit|core binding factor subunit beta|Core-binding factor subunit beta|Core-binding factor, beta subunit (CBFB), transcript variant 2|PEA 2|PEA2|PEA2 beta|PEA2-beta|PEA2beta|PEBB_HUMAN|PEBP 2B|PEBP2 beta|PEBP2-beta|PEBP2B|PEBP2beta|Polyomavirus enhancer binding protein 2 beta subunit|Polyomavirus enhancer-binding protein 2 beta subunit|SL3 3 enhancer factor 1 beta subunit|SL3 3 enhancer factor 1 subunit beta|SL3-3 enhancer factor 1 subunit beta|SL3/AKV core binding factor beta subunit|SL3/AKV core-binding factor beta subunit
- CBF b|CBF beta|CBF-beta|CBFB|CBFbeta|Core binding factor beta subunit|core binding factor subunit beta|Core-binding factor subunit beta|Core-binding factor, beta subunit (CBFB), transcript variant 2|PEA 2|PEA2|PEA2 beta|PEA2-beta|PEA2beta|PEBB_HUMAN|PEBP 2B|PEBP2 beta|PEBP2-beta|PEBP2B|PEBP2beta|Polyomavirus enhancer binding protein 2 beta subunit|Polyomavirus enhancer-binding protein 2 beta subunit|SL3 3 enhancer factor 1 beta subunit|SL3 3 enhancer factor 1 subunit beta|SL3-3 enhancer factor 1 subunit beta|SL3/AKV core binding factor beta subunit|SL3/AKV core-binding factor beta subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13951
- Q13951
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together