Anti-ZC3HC1 Rabbit Polyclonal Antibody
77804-909
New Product
- Antigen name:Zinc finger C3HC-type protein 1
- Antigen symbol:ZC3HC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q86WB0
- Antigen synonyms:hNIPA|NIPA_HUMAN|Nuclear interacting partner of ALK|Nuclear interacting partner of anaplastic lymphoma kinase|Nuclear-interacting partner of ALK|Nuclear-interacting partner of anaplastic lymphoma kinase|zc3hc1|Zinc finger C3HC type containing 1|Zinc finger C3HC-type protein 1
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: ZC3HC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat