Anti-RND2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB:1:500 to 1:5000
Type:
Antigen: RND2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
- Catalog No:
- 77797-422
- 77797-421
- Antigen symbol:
- RND2
- RND2
- Antigen name:
- Rho-related GTP-binding protein RhoN protein (104-163AA)
- Rho-related GTP-binding protein RhoN protein (104-163AA)
- Antigen synonyms:
- ARHN|CTD-3199J23.4|GTP-binding protein Rho7|ras homolog gene family, member N|Rho 7|Rho family GTPase 2|Rho N|Rho related GTP-binding protein RhoN|Rho-related GTP-binding protein Rho7|Rho-related GTP-binding protein RhoN|Rho7|RhoN|Rnd 2|Rnd2|RND2_HUMAN
- ARHN|CTD-3199J23.4|GTP-binding protein Rho7|ras homolog gene family, member N|Rho 7|Rho family GTPase 2|Rho N|Rho related GTP-binding protein RhoN|Rho-related GTP-binding protein Rho7|Rho-related GTP-binding protein RhoN|Rho7|RhoN|Rnd 2|Rnd2|RND2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52198
- P52198
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together